Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CBLN4 Recombinant Protein

Catalog No. 89015798 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-015-798 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-015-798 Supplier Abnova™ Supplier No. H00140689P01
Add to Cart
Add to Cart

Human CBLN4 full-length ORF recombinant protein with GST-tag at N-terminal

Cerebellin is a sixteen aa peptide found mainly in the adrenal medulla, where it has been shown to have a neuromodulatory function. Cerebellin is derived from precerebellin, a protein with sequence similarity to the noncollagen domain of complement component C1qB. The protein encoded by this gene is a glycoprotein which shares sequence similarity with precerebellin.

  • Theoretical MW (kDa): 48.2
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MGSGRRALSAVPAVLLVLTLPGLPVWAQNDTEPIVLEGKCLVVCDSNPATDSKGSSSSPLGISVRAANSKVAFSAVRSTNHEPSEMSNKTRIIYFDQILVNVGNFFTLESVFVAPRKGIYSFSFHVIKVYQSQTIQVNLMLNGKPVISAFAGDKDVTREAATNGVLLYLDKEDKVYLKLEKGNLVGGWQYSTFSGFLVFPL

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Molecular Weight (g/mol) 48.2
Name Human CBLN4 Full-length ORF Recombinant Protein with GST-tag at N-terminal
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE stained with Coomassie Blue
Quantity 10 μg
Source Wheat Germ (in vitro)
Storage Requirements -80°C
Cross Reactivity Human
Recombinant Recombinant
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.