Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBP Monoclonal Antibody (NM11)
GREENER_CHOICE

Catalog No. PIMA513634
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513634 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513634 Supplier Invitrogen™ Supplier No. MA513634
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-13634 targets p300/CBP (CREB-Binding Protein) in ICC/IF and IHC (P) applications and shows reactivity with Human, Mink, mouse, Non-human primate, and Rat samples. The MA5-13634 immunogen is e1A affinity purified p300 protein.

CBP for CREB-binding protein is a 265 kDa nuclear protein which probably has many of the properties expected of a CREB co-activator. It shows substantial sequence homology with p300.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBP
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone NM11
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene CREBBP
Gene Accession No. P45481, Q6JHU9, Q92793
Gene Alias AW558298; CBP; CBP/p300; CREB binding protein; CREB-binding protein; CREBBP; KAT3A; p300/CBP; RSTS; RTS
Gene Symbols CREBBP
Host Species Mouse
Immunogen E1A affinity purified p300 protein.
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12914, 1387, 54244
Target Species Human, Mink, Mouse, Monkey, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.