Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ CBR 2
SDPSCI_ED

Catalog No. S15988ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Application):
Educational use only
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S15988ND Educational use only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15988ND Supplier Vernier™ Supplier No. CBR2

missing translation for 'validMainframeMsg'

Includes

Texas Instruments Calculator-Based Ranger™ 2, CBR 2 to Calculator Cable, CBR 2 to TI-Nspire Cable, User manual

Requires

No interface is needed. Data-collection software included with compatible TI handhelds.

The least expensive and easiest way to collect motion data in your math, physical science, or physics classroom is with the CBR 2.

  • No interface required
  • The measurement range is 0.15 m to 6 m.
  • CBR 2 has a sensitivity switch which lets you customize settings to your experiment.
  • The CBR 2 is built with a pivoting head, making it easy to set up experiments.
  • It works with TI-73 Explorer, TI-82, TI-83, TI-83 Plus, TI-83 Plus Silver Edition, TI-84 Plus, TI-84 Plus Silver Edition, TI-84 Plus C Silver Edition, TI-84 Plus CE, TI-86, TI-89, TI-92, TI-92 Plus, TI-Nspire, TI-Nspire CX, TI-Nspire CAS, TI-Nspire CAS CX, and TI Voyage 200 calculators.
  • The CBR 2 can also be connected to a CBL, CBL 2, LabPro, LabQuest, or LabQuest Mini (using MDC-BTD cable, sold separately).
  • Use CBR 2 to teach important concepts in physics, physical science, and math.
  • Engage your students with hand-on activities.
  • Study the motion of a ball tossed in the air, or a cart on a ramp.
  • Attach CBR 2 directly to the Vernier Dynamics Track.
  • The least expensive and easiest way to collect motion data in your math, physical science, or physics classroom is with the CBR 2.

Warranty and Services

1 year warranty from Texas Instruments

Specifications

Includes CBR 2 to Calculator Cable, CBR 2 to TI-Nspire Cable, User Manual
Grade Middle School and High School
Warranty 1-year limited warranty from Texas Instruments
For Use With (Application) Educational use only
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.