Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBR3 Recombinant Rabbit Monoclonal Antibody (K01_1P02)

Catalog No. MA564674
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA564674 50 μL
MA563215 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA564674 Supplier Invitrogen™ Supplier No. MA564674
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

CBR3 Recombinant Monoclonal Antibody for Western Blot

Carbonyl reductase 3 catalyzes the reduction of a large number of biologically and pharmacologically active carbonyl compounds to their corresponding alcohols. The enzyme is classified as a monomeric NADPH-dependent oxidoreductase. CBR3 contains three exons spanning 11.2 kilobases and is closely linked to another carbonyl reductase gene - CBR1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBR3
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1P02
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene CBR3
Gene Accession No. O75828
Gene Alias 1110001J05Rik; C81353; carbonyl reductase (NADPH) 3; carbonyl reductase [NADPH] 3; carbonyl reductase 3; Cbr3; epididymis secretory protein Li 25; hCBR3; HEL-S-25; NADPH-dependent carbonyl reductase 3; SDR21C2; Short chain dehydrogenase/reductase family 21C member 2; short chain dehydrogenase/reductase family 21C, member 2
Gene Symbols CBR3
Host Species Rabbit
Immunogen A synthetic peptide of human CBR3.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 874
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.