Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBF Bioscience Electrophoretic Blotting System: EBU-102 (Three Gels)
SCI_ED

Catalog No. S05778 Shop All CBS Scientific Products
Change view
Click to view available options
EBU-102; three gels
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S05778 EBU-102; three gels
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05778 Supplier MBF Bioscience Supplier No. EBU102

missing translation for 'validMainframeMsg'

Blotters for mini gels

  • For Western, Southern, Northern blots
  • 3 Gel cassette capacity
  • Transfer gels as large as 16 x 22cm
  • Tank blotter includes platinum wire electrode panels, internal cooling base, 1 gel cassette and safety cover with attached power leads

Certifications

CE marked

Specifications

Certifications/Compliance CE
Includes Blotting chamber with room for spin bar beneath cassettes and internal cooling base for temperature controlled runs, 2 Platinum electrode panels, 1 blotting cassette with 2 sponge pads, safety cover with power Leads, tubing adapters
Volume (English) Buffer 127.47 oz.
Volume (Metric) Buffer 3770 mL
Dimensions (L x W x H) 7.48 x 4.3 x 11.22 in. (19 x 11 x 28.5 cm)
Length (English) 7.48 in.
Length (English) Gel 8.66 in.
Length (Metric) 19 cm
Length (Metric) Gel 22 cm
Width (English) 4.3 in.
Width (English) Gel 6.29 in.
Width (Metric) 11 cm
Width (Metric) Gel 16 cm
Height (English) 11.22 in.
Height (English) Gel 8.66 in.
Height (Metric) 28.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.