Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBF Bioscience QNX-700 Quadra Mini-Vertical PAGE/Blotting System
SCI_ED

Catalog No. S05769 Shop All CBS Scientific Products
Change view
Click to view available options
Ambient Temperature (English):
140°F
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Ambient Temperature (English)
S05769 140°F
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05769 Supplier MBF Bioscience Supplier No. QNX700HC

missing translation for 'validMainframeMsg'

Quadruple Capacity with Dual Function - 4 slab gels or 4 electrophoretic transfers can be accomplished in the same unit.

  • Will accept 10cm(w) x 10cm(l), 10cm(w) x 9cm(l) or 10cm(w) x 8cm(l) precast gels
  • Gel or blotting cassettes can be extracted from the core without removing the core from the tank, allowing for quick processing of samples
  • Allows for the immediate reuse of buffer for the next set of samples
  • Hinged core design allows easy loading and removal of gel or blotting cassettes
  • Color-coded anode and cathode safety features on all components
  • Tank design includes stirring bar corral
  • Uses 950mls-1400mls of buffer for either application
  • There are no loose parts to keep track of on the central core

  • Designed for use with ClearPAGE or other Precast gels
  • Handcasting can be accomplished using Gel Wrap Casting kits and Multiple Gel Casting Chamber kits
  • Without internal cooling
  • Can accommodate both slab gel electrophoresis and Western blotting within the same unit without the purchase of additional modules
  • Equipped with: Lower buffer reservoir, two cores to accommodate a total of four gels or four blotting cassettes, four blotting cassettes with sponge pads, Safety cover with attached power leads

SDS-PAGE, Electro-Blotting (Western), Acrylamide nucleic acid separations

Certifications

CE marked

Specifications

Ambient Temperature (English) 140°F
Ambient Temperature (Metric) 60°C
Certifications/Compliance EN61010-1-1993 (IEC1010-1), CE
Device Holds Four 10 x 10cm precast Gel or handcast Gel
Type Quadra Mini Vertical PAGE/Blotting System
Includes 4 Place Gel Wrap™ hand casting bundle (4 combs, 4 sets spacers, 4 Glass plate sets and 4 Gel Wrap™ gaskets, 12 clamps), lower Buffer reservoir, 2 cores to accommodate 4 gels or 4 blotting cassettes, 4 blotting cassettes
Volume (English) Buffer 32.12 to 37.19 oz.
Volume (Metric) Buffer 950 to 1100 mL
Dimensions (L x W x H) 17.1 x 17.8 x 15.2 in. (43.4 x 45.2 x 38.6 cm)
Length (English) 17.1 in.
Length (English) Gel 3.93, 3.54, 3.14 in.
Length (Metric) 43.4 cm
Length (Metric) Gel 10, 9, 8 cm
Width (English) 17.8 in.
Width (English) Gel 3.93 in.
Width (Metric) 45.2 cm
Width (Metric) Gel 10 cm
Height (English) 15.2 in.
Height (Metric) 38.6 cm
Voltage 300 V
Wattage 30 w
Amperage 190 mA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.