Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBF Bioscience Vertical Gel Kits
SCI_ED

Catalog No. S05774 Shop All CBS Scientific Products
Change view
Click to view available options
Lite Slab Gel Kit; 14.5cm pathlength
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S05774 Lite Slab Gel Kit; 14.5cm pathlength
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05774 Supplier MBF Bioscience Supplier No. WSP2SG125

missing translation for 'validMainframeMsg'

Includes

2 combs, 2 sets glass plates, 2 sets of spacers, 2 gel wrap gaskets, 8 white spring clamps

Features leveling base, platinum electrodes and safety-tip power leads.

  • Long-lasting silicone sponge gasket on upper reservoir prevents buffer leakage
  • 16.5cm width with two sets accessories

Separating proteins, SDS PAGE

Specifications

Includes 2 Combs, 2 sets Glass plates, spacers, gasket, 8 spring clamps
Width (English) 6.49 in.
Width (Metric) 16.5 cm
Height (English) 5.7 in.
Height (Metric) 14.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.