Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBX2 Chimeric Recombinant Rabbit Monoclonal Antibody (RAB-C238)
GREENER_CHOICE

Catalog No. PIMA548333 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548333 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548333 Supplier Invitrogen™ Supplier No. MA548333
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Specificity: This antibody recognizes CBX2 (Chromobox protein homolog 2). It binds to a folded domain, amino acids 2-70. CBX2 is a component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development.

MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBX2 Chimeric
Applications ChIP Assay, ELISA, Flow Cytometry, Immunoprecipitation, Immunocytochemistry
Classification Recombinant Monoclonal
Clone RAB-C238
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene CBX2
Gene Accession No. Q14781
Gene Alias Cbx2; CDCA6; cell division cycle associated 6; chromobox 2; chromobox homolog 2; chromobox homolog 2 (Pc class homolog, Drosophila); chromobox protein homolog 2; hromobox homolog 2; M33; M33 polycomb-like protein; MOD2; modifier 3; modifier 3 protein; pc; Pc class homolog; polycomb; SRXY5
Gene Symbols CBX2
Host Species Rabbit
Immunogen This antibody was obtained by recombinant antibody (rAb) phage display recognizing CBX2 protein under non-denaturing conditions.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84733
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.