Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBX3 Mouse anti-Human, Mouse, Clone: 1A5A3, Proteintech
SDP

Catalog No. p-7212722 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6872 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6872 Supplier Proteintech Group Inc Supplier No. 664461IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

At the nuclear envelope, the nuclear lamina and heterochromatin are adjacent to the inner nuclear membrane. The protein encoded by this gene binds DNA and is a component of heterochromatin. This protein also can bind lamin B receptor, an integral membrane protein found in the inner nuclear membrane. The dual binding functions of the encoded protein may explain the association of heterochromatin with the inner nuclear membrane.

Specifications

Antigen CBX3
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1A5A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CBX3
Gene Accession No. P23198, Q13185
Gene Alias CBX3, Chromobox protein homolog 3, HECH, HP1 GAMMA, HP1Hs gamma, Modifier 2 protein
Gene Symbols CBX3
Host Species Mouse
Immunogen CBX3 Fusion Protein Ag19112
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11335, 12417
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.