Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBX8 Monoclonal Antibody (8G7)
GREENER_CHOICE

Catalog No. PIMA549285
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549285 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549285 Supplier Invitrogen™ Supplier No. MA549285
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human A549 whole cell, human Raji whole cell. IHC: human gallbladder adenocarcinoma tissue. Flow: HL-60 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBX8
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 8G7
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene CBX8
Gene Accession No. Q9HC52
Gene Alias CBX8; chromobox 8; chromobox homolog 8; chromobox homolog 8 (Pc class homolog, Drosophila); chromobox protein homolog 8; HPC3; mPc3; Pc class 3 homolog; Pc3; polycomb 3; polycomb 3 homolog; RC1; rectachrome 1
Gene Symbols CBX8
Host Species Mouse
Immunogen E.coli-derived human Cbx8 recombinant protein (Position: M1-R389).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57332
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.