Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBX8 Recombinant Rabbit Monoclonal Antibody (4H2L5)

Catalog No. 703495
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-349-5 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-349-5 Supplier Invitrogen™ Supplier No. 703495
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Mouse, Rat, Bovine.

Component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development. PcG PRC1 complex acts via chromatin remodeling and modification of histones; it mediates monoubiquitination of histone H2A 'Lys-119', rendering chromatin heritably changed in its expressibility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBX8
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 4H2L5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene CBX8
Gene Accession No. Q9HC52
Gene Alias CBX8; chromobox 8; chromobox homolog 8; chromobox homolog 8 (Pc class homolog, Drosophila); chromobox protein homolog 8; HPC3; mPc3; Pc class 3 homolog; Pc3; polycomb 3; polycomb 3 homolog; RC1; rectachrome 1
Gene Symbols CBX8
Host Species Rabbit
Immunogen Protein corresponding to human CBX8 [aa81-aa281].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57332
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.