Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CBY1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550348
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550348 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550348 Supplier Invitrogen™ Supplier No. PA550348
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total CBY1 protein.

Beta-catenin is a transcriptional activator and oncoprotein involved in the development of several cancers. The protein encoded by this gene interacts directly with the C-terminal region of beta-catenin, inhibiting oncogenic beta-catenin-mediated transcriptional activation by competing with transcription factors for binding to beta-catenin. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CBY1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CBY1
Gene Accession No. Q9Y3M2
Gene Alias 1110014P06Rik; arb1; ARPP-binding protein; C22orf2; CBY; CBY1; chibby CTNNB1-mediated transcription inhibitor; chibby family member 1, beta catenin antagonist; chibby homolog 1; chibby homolog 1 (Drosophila); coiled-coil protein PIGEA-14; cytosolic leucine-rich protein; HRIHFB2025; HS508I15A; PGEA1; PGEA14; PIGEA14; PIGEA-14; PKD2 interactor, golgi and endoplasmic reticulum associated 1; PKD2 interactor, Golgi and endoplasmic reticulum-associated 1; polycystin-2 interactor, Golgi- and endoplasmic reticulum-associated protein, 14 kDa; protein chibby homolog 1
Gene Symbols CBY1
Host Species Rabbit
Immunogen Full length fusion protein of Human CBY1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25776
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.