Learn More
Medchemexpress LLC HY-138304 5mg Medchemexpress, CC-90001 CAS:1403859-14-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1383045MG
Encompass_Preferred
Medchemexpress, HY-138304 5mg CC-90001 CAS:1403859-14-2 CC-90001 is a potent, selective and orally active inhibitor of c-Jun N-terminal kinase (JNK). CC-90001 shows 12.9-fold selectivity for JNK1 over JNK2 in a cell-based model. CC-90001 can be used for the research of idiopathic pulmonary fibrosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1522