Learn More
Medchemexpress LLC CC-90002 1mg | 1mg
Supplier: Medchemexpress LLC HYP991721MG
CC-90002 is a humanized anti-CD47 monoclonal antibody (mAb) CC-90002 has a high affinity for binding to CD47 with a subnanomolar Kd value CC-90002 can be used for the research of hematologic malignancies and solid tumors[1][2]
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.