Learn More
Medchemexpress LLC HY-137573 5mg Medchemexpress, CC-90010 CAS:1706738-98-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1375735MG
Encompass_Preferred
Medchemexpress, HY-137573 5mg CC-90010 CAS:1706738-98-8 CC-90010 (compound 1) is a reversible and orally active BET inhibitor. CC-90010 is applied in the study for advanced solid tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1502
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.