Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CC2D1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7210027 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3076 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3076 Supplier Proteintech Group Inc Supplier No. 207741AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is specific to CC2D1B, which has been confirmed by siRNA test.

Transcription factor that binds specifically to the DRE (dual repressor element) and represses HTR1A gene transcription in neuronal cells.

Specifications

Antigen CC2D1B
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CC2D1B
Gene Accession No. Q5FVK6, Q5T0F9, Q8BRN9
Gene Alias CC2D1B, Freud-2, KIAA1836, LGD1
Gene Symbols Cc2d1b
Host Species Rabbit
Immunogen CC2D1B Fusion Protein Ag14767
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200014, 313478, 319965
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.