Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC111 Monoclonal Antibody (10-B3)
GREENER_CHOICE

Catalog No. PIMA532899
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532899 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532899 Supplier Invitrogen™ Supplier No. MA532899
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCDC111 Monoclonal Antibody for ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC111
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone 10-B3
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene PRIMPOL
Gene Accession No. Q96LW4
Gene Alias CCDC111; coiled-coil domain containing 111; coiled-coil domain-containing protein 111; DNA-directed primase/polymerase protein; hPrimpol1; MYP22; primase and DNA directed polymerase; primase and polymerase (DNA-directed); PRIMPOL
Gene Symbols PRIMPOL
Host Species Mouse
Immunogen Recombinant protein within Human CCDC111 aa 1-560.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201973
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.