Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC132 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555374
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555374 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555374 Supplier Invitrogen™ Supplier No. PA555374
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSRSPSVSPS KQPVSTSSKT VTLFEQYCSG GNPFEIQANH KDEETEDVLA SNGYESDEQE KSAYQEYDSD SDVPEELKRD YVDEQTGDGP VKSVSRETLK SRKKSDYSLN KVNAP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

CCDC132 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC132
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Vps50
Gene Accession No. F1LSG8, Q8CI71, Q96JG6
Gene Alias 1700034M03Rik; 8430415E05Rik; Bcl2-like protein blm; BLM; C87205; CCDC132; coiled-coil domain containing 132; coiled-coil domain-containing protein 132; DKFZp313I2429; EARP/GARPII complex subunit VPS50; FLJ20097; KIAA1861; mKIAA1861; RGD1304751; syndetin; Vps50; VPS50 EARP/GARPII complex subunit; VPS50, EARP/GARPII complex subunit; VPS54L
Gene Symbols Vps50
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC132. Recombinant protein control fragment (Product #RP-92587).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 312083, 55610, 73288
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.