Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC16 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560339
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
50-156-0339 100 μL
1 options

Catalog No. 50-156-0339

Supplier: Bethyl Laboratories, Inc A301419A

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat

Located on chromosome 17, this gene encodes for a coiled-coil domain containing 16 protein with unknown function.

Specifications

Antigen CCDC16
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ZNF830
Gene Accession No. Q96NB3
Gene Alias CCDC16, coiled-coil domain containing 16, coiled-coil domain-containing protein 16, hypothetical protein MGC20398, MGC20398, OMCG1, orphan maintece of genome 1
Gene Symbols ZNF830
Host Species Rabbit
Immunogen Between 322 and 372
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91603
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.