Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCDC181 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555579
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555579 Supplier Invitrogen™ Supplier No. PA555579
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSNHRTQSAH ISPVTSTYCL SPRQKELQKQ LEEKREKLKR EEERRKIEEE KEKKRENDIV FKAWLQKKRE QVLEMRRIQR AK Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CCDC181
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC181
Gene Accession No. Q5TID7, Q80ZU5
Gene Alias 4930455F23Rik; AI427889; C1orf114; Ccdc181; coiled-coil domain containing 181; coiled-coil domain-containing protein 181; RGD1309708
Gene Symbols CCDC181
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CCDC181. Recombinant protein control fragment (Product #RP-94119).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57821, 74895
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.