Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCF2-FA (free acid)

CCF2-FA (free acid)

$844.00 - $6360.00

Specifications

Cell Permeability Cell-Impermeant
Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Product Line GeneBLAzer™
Substrate CCF2-FA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
K1027
Encompass_Preferred
View Documents
LIFE TECHNOLOGIES
K1027
1mg
Each for $6,360.00
Only null left
Add to Cart
 
K1034
Encompass_Preferred
View Documents
LIFE TECHNOLOGIES
K1034
100μg
Each for $844.00
Only null left
Edge
Add to Cart
 
Description

Description

CCF2-FA (Free Acid) is a fluorescence resonance energy transfer (FRET) substrate that consists of a cephalosporin core linking B7-hydroxycoumarin to fluorescein. This product is intended for use as a beta-lactamase substrate in a GeneBLAzer™ cell-based assay.

GeneBLAzer™ cell-based assays typically utilize the membrane-permeant ester forms (CCF2-AM and CCF4-AM) of the negatively charged fluorescent beta-lactamase substrates, CCF2 and CCF4. These lipophilic esters readily enter the cell, where cleavage by endogeneous cytoplasmic esterases rapidly converts them into their negatively charged forms, thereby trapping them in the cytosol.

CCF2-FA is essentially the CCF2 substrate without the esters found in the AM version. CCF2-FA is de-esterified and used in cell lysate applications, bypassing loading across the cell membrane and de-esterification steps. Cell lysates are the preferred method for applications using cells that contain a cell wall. CCF2-FA can also be used as a control to acquire the excitation and emission spectra for CCF2-AM and CCF4-AM.
Specifications

Specifications

Cell-Impermeant
Fluorescence
CCF2-FA
Beta-Lactamase Substrate
Store in freezer (-5 to -30°C).
GeneBLAzer™
Chemical Substrate
Approved for shipment on Wet or Dry Ice
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.