Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCK Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501728468 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8468 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8468 Supplier Proteintech Group Inc Supplier No. 130742AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cholecystokinin (CCK) is a neuropeptide hormone and neurotransmitter widely distributed throughout the gastrointestinal (GI) tract and the central nervous system (CNS). Antibodies that react specifically with CCK are useful for the study of the mode of action, differential tissue expression and intracellular and subcellular localization of CCK in the CNS and periphery as well as in neuroendocrine cells of the digestive system.

Specifications

Antigen CCK
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CCK
Gene Accession No. P06307
Gene Alias CCK, CCK12 Cholecystokinin 8, CCK18 Cholecystokinin 12, CCK25 Cholecystokinin 18, CCK33 Cholecystokinin 25, CCK39 Cholecystokinin 33, CCK5, CCK7 Cholecystokinin 5, CCK8 Cholecystokinin 7, cholecystokinin
Gene Symbols CCK
Host Species Rabbit
Immunogen CCK Fusion Protein Ag3725
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 885
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.