Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCKAR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA3116 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA3116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA3116 Supplier Invitrogen™ Supplier No. PA3116
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA3-116 detects CCK1-Receptor in human samples. PA3-116 has been successfully used in Western blot, immunocytochemistry and immunohistochemistry procedures. By Western blot, this antibody detects an ~80-90 kDa protein representing CCK1-Receptor in human samples. In immunohistochemistry, PAC1 immunoreactivity is predominantly localized at the plasma membrane and in cells of the gastric mucosa, myenteric ganglion cells, myenteric nerve fibers in the gasrtointestinal tract. PA3-116 immunogen is a synthetic peptide, KLH conjugated, corresponding to the residues T(409) G A S L S R F S Y S H M S A S V P P Q (428) of human VPAC2 receptor.

This gene encodes a G-protein coupled receptor that binds non-sulfated members of the cholecystokinin family of peptide hormones. This receptor is a major physiologic mediator of pancreatic enzyme secretion and smooth muscle contraction of the gallbladder and stomach. In the central and peripheral nervous system this receptor regulates satiety and the release of beta-endorphin and dopamine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCKAR
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole Serum with 0.05% sodium azide
Gene CCKAR
Gene Accession No. P32238
Gene Alias AW106902; CCK1R; CCK1-R; CCK-A; ccka receptor; CCK-A receptor; Cckar; CCK-AR; CCKRA; cholecystokinin A receptor; cholecystokinin receptor type A; cholecystokinin type-A receptor; cholecystokinin-1 receptor; HGNC:1570
Gene Symbols CCKAR
Host Species Rabbit
Immunogen Synthetic peptide, KLH conjugated, corresponding to the residues T(409) G A S L S R F S Y S H M S A S V P P Q (428) of human CCK1 receptor.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 886
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.