Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCL14 Recombinant Rabbit Monoclonal Antibody (007)
GREENER_CHOICE

Catalog No. PIMA530136
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530136 Supplier Invitrogen™ Supplier No. MA530136
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human CCL14.

This gene, CCL14, is one of several CC cytokine genes clustered on 17q11.2. The CC cytokines are secreted proteins characterized by two adjacent cysteines. The cytokine encoded by this gene induces changes in intracellular calcium concentration and enzyme release in monocytes. This gene expresses both monocistronic and bicistronic transcripts. Bicistronic transcripts include the upstream cytokine gene CCL15. In addition, CCL14 undergoes alternative splicing in the bicistronic transcripts; it is unknown if its monocistronic transcript is also subject to alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL14
Applications ELISA
Classification Recombinant Monoclonal
Clone 007
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CCL14
Gene Accession No. Q16627
Gene Alias C-C motif chemokine 14; C-C motif chemokine ligand 14; CC-1; CC-3; CCL14; chemokine (C-C motif) ligand 14; chemokine CC-1/CC-3; chemokine CC-3; CKB1; H HCC-1(1-74); HCC-1; HCC-1(1-74); HCC-1(3-74); HCC-1(4-74); HCC-1(9-74); HCC-1/HCC-3; HCC-3; hemofiltrate CC chemokine 1; MCIF; NCC2; NCC-2; new CC chemokine 2; SCYA14; SCYL2; small inducible cytokine subfamily A (Cys-Cys), member 14; small-inducible cytokine A14; SY14
Gene Symbols CCL14
Host Species Rabbit
Immunogen Recombinant Human CCL14 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6358
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.