Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CCL23/Ck beta 8-1/MIP3 Antibody Pair [HRP]
SDP

Catalog No. NB010041 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010041 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010041 Supplier Novus Biologicals™ Supplier No. NBP27950115PLATES
Add to Cart
Add to Cart

Polyclonal/Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 6.25 pg/mL
Assay Sensitivity 6.25-400 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Human CCL23/Ck beta 8-1/MIP3.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias C6 beta-chemokine, chemokine (C-C motif) ligand 23, CKb8, CKB-8, CK-beta-8, hmrp-2a, Macrophage inflammatory protein 3, MIP3, MIP-3CK-BETA-8, MPIF1, MPIF-1Ckb-8-1, Myeloid progenitor inhibitory factor 1, SCYA23C-C motif chemokine 23, small inducible cytokine subfamily A (Cys-Cys), member 23, Small-inducible cytokine A23
Gene ID (Entrez) 6368
Gene Symbol CCL23
Host Species Rabbit
Kit Contents
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human CCL23/Ck beta 8-1/MIP3 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1 mg/mL of rabbit anti-Human CCL23/Ck beta 8-1/MIP3 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 10 ng of recombinant Human CCL23/Ck beta 8-1/MIP3.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Biologically Active Proteins, Cell Cycle and Replication
    Storage Requirements Storage is content dependent.
    Target CCL23/Ck beta 8-1/MIP3
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 400 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.