Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech CCL3L1 Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. 500P187G50
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P187G50 50 μg
500P187G100 100 μg
500P187G1MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 500P187G50 Supplier PeproTech Supplier No. 500P187G50UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

AA Sequence of recombinant protein: APLAADTPTA CCFSYTSRQI PQNFIADYFE TSSQCSKPSV IFLTKRGRQV CADPSEEWVQ KYVSDLELSA. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human LD78beta (CCL3L1). Anti-Human LD78beta (CCL3L1)-specific antibody was purified by affinity chromatography employing an immobilized Human LD78beta (CCL3L1) matrix. Sandwich ELISA: To detect Human LD78beta (CCL3L1) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human LD78beta (CCL3L1) (500-P187GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human LD78beta (CCL3L1). Western Blot: To detect Human LD78beta (CCL3L1) by Western Blot analysis this antibody can be used at a concentration of 0.1- 0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human LD78beta (CCL3L1) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

This gene is one of several cytokine genes that are clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins that function in inflammatory and immunoregulatory processes. The protein encoded by this gene binds to several chemokine receptors, including chemokine binding protein 2 and chemokine (C-C motif) receptor 5 (CCR5). CCR5 is a co-receptor for HIV, and binding of this protein to CCR5 inhibits HIV entry. The copy number of this gene varies among individuals, where most individuals have one to six copies, and a minority of individuals have zero or more than six copies. There are conflicting reports about copy number variation of this gene and its correlation to disease susceptibility. This record represents one of two copies that are present on the ALT_REF_LOCI_2 alternate haplotype of the GRCh38 human reference genome assembly. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCL3L1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CCL3L1
Gene Accession No. P16619
Gene Alias 464.2; C-C motif chemokine 3-like 1; C-C motif chemokine ligand 3 like 1; CC-chemokine (CCL3); MIP1a; CCL3L1; CCL3L3; chemokine (C-C motif) ligand 3-like 1; chemokine ligand 3-like 1; D17S1718; G0/G1 switch regulatory protein 19-2; G0S19-2; LD78; LD78BETA; LD78-beta(1-70); LD78-beta(3-70); LD78-beta(5-70); macrophage inflamatory protein 1 alpha; MIP1AP; PAT 464.2; SCYA3L; SCYA3L1; small inducible cytokine A3-like 1; Small-inducible cytokine A3-like 1; Tonsillar lymphocyte LD78 beta protein
Gene Symbols CCL3L1
Host Species Goat
Immunogen E.coli-derived Recombinant Human LD78beta (CCL3L1).
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6349
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.