Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCNH Antibody, ProSci
SDP

Catalog No. 16830220
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-830-220 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-830-220 Supplier ProSci LLC Supplier No. 310350100
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCNH Polyclonal Antibody specifically detects CCNH in Human samples. It is validated for ELISA, Western Blot

Specifications

Antigen CCNH
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution CCNH antibody can be used for detection of CCNH by ELISA at 1:75000. CCNH antibody can be used for detection of CCNH by western blot at 1 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Formulation Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Gene Accession No. NP_001230
Gene Alias CCNH, CAK, p34, p37
Host Species Rabbit
Immunogen Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human CCNH.
Purification Method Antibody is purified by Protein A/G affinity chromatography method.
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 902
Target Species Human
Content And Storage For short periods of storage (days) store at 4°C. For longer periods of storage, store CCNH antibody at -20°C. As with any antibody avoid repeat freeze-thaw cycles.
Form Liquid
Research Category Cell Cycle, Cancer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.