Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCNK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116511
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116511 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116511 Supplier Invitrogen™ Supplier No. PA5116511
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, Mouse brain Immunogen sequence: MKENKENSSP SVTSANLDHT KPCWYWDKKD LAHTPSQLEG LDPATEARYR REGARFIFDV GTRLGLHYDT LATGIIYFHR FYMFHSFKQF PRYVTGACCL FLAGKVEETP KKCKDIIKTA RSLLNDVQFG QFGDDPKEEV MVLERILLQT IKFDLQVEHP.

Regulatory subunit of cyclin-dependent kinases that mediates activation of target kinases. Plays a role in transcriptional regulation via its role in regulating the phosphorylation of the C-terminal domain (CTD) of the large subunit of RNA polymerase II (POLR2A).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCNK
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CCNK
Gene Accession No. O75909, O88874
Gene Alias AW123198; AW413594; CCNK; CPR4; CycK; cyclin K; cyclin-K
Gene Symbols CCNK
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human Cyclin K (NP_0010928721).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12454, 500715, 8812
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.