Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113003
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113003 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113003 Supplier Invitrogen™ Supplier No. PA5113003
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCP2 Polyclonal Antibody for IHC (P), ELISA

Metallocarboxypeptidase that mediates deglutamylation of target proteins. Catalyzes the deglutamylation of polyglutamate side chains generated by post-translational polyglutamylation in proteins such as tubulins. Also removes gene-encoded polyglutamates from the carboxy-terminus of target proteins such as MYLK. Does not show detyrosinase or deglycylase activities from the carboxy-terminus of tubulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCP2
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene AGBL2
Gene Accession No. Q5U5Z8
Gene Alias 4930524K04; A430081C19Rik; Agbl2; ATP/GTP binding protein like 2; ATP/GTP binding protein-like 2; ATP/GTP-binding protein-like 2; carboxypeptidase 2, cytosolic; CBCP2; CCP2; cytoplasmic carboxypeptidase 2; cytosolic carboxypeptidase 2; testis tissue sperm-binding protein Li 96mP; testis-expressed protein 25; zebrafish testis-expressed 25; zgc:165648; zte25
Gene Symbols AGBL2
Host Species Rabbit
Immunogen Recombinant Human Cytosolic carboxypeptidase 2 protein (10-270AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79841
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.