Learn More
IBL America CCPlus: Anti-Cyclic Citrullinated
Supplier:  IBL America RA96PLUS
Encompass
The Immunoscan CCPlus test kit is an enzyme-linked immunosorbent assay (ELISA) for the determination of IgG antibodies to Cyclic Citrullinated Peptides (CCP) in human serum. 510(k) cleared - for in vitro diagnostic use only.
Catalog No. NC1831091
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.