Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR3 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501733844 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3844 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3844 Supplier Proteintech Group Inc Supplier No. 223511AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Chemokines play important roles in inflammation and critical for the recruitment of effector immune cells to sites of infection. Chemokines activate leukocytes by binding to G protein coupled receptors. The ever-growing chemokine receptor subtypes can be divided into 2 major groups, CXCR and CCR, based on the 2 major classes of chemokines. One of the CCR receptors, CCR3 (eotaxin receptor), is expressed on eosinophils and certain T cell population respond to a variety of CC chemokines apart from eotaxin, including RANTES, monocyte chemotactic protein (MCP)-2, MCP-3, and MCP-4. CCR3 facilitated infection by a more restricted subset of primary viruses, and binding of the CCR3 ligand, eotaxin, and inhibited infection by these isolates.

Specifications

Antigen CCR3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CCR3
Gene Accession No. P51677
Gene Alias C C chemokine receptor type 3, C C CKR 3, CC CKR 3, CCR 3, CCR3, CD193, CKR3, CMKBR3, Eosinophil eotaxin receptor
Gene Symbols CCR3
Host Species Rabbit
Immunogen CCR3 Fusion Protein Ag17926
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1232
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.