Learn More
Selleck Chemical LLC CCT245737 5mg
SDP
Supplier:  Selleck Chemical LLC S82535MG
Encompass_Preferred
CCT245737 (SRA737, PNT-737) is an orally active CHK1 inhibitor with The IC50 of 1.4 nM. It exhibits >1,000-fold selectivity against CHK2 and CDK1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3350
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.