Learn More
Julabo USA Inc CORIO CD-600F refrigerated/heating circulator, 1kW heater, 600W cooling power, 115V/60Hz
SDP
Supplier:  Julabo USA Inc 9012704.02
Encompass_Preferred
CORIO™ CD-600F refrigerated/heating circulator. Suitable for temperature applications with circulation to an external apparatus or immersing a device in the bath tank. The CORIO™ CD circulator incorporates easy change-over for control of fluid circulation internally or externally. Operating range from -35…+150 °C with 1kW heating power, 600W cooling power. Unit fittings M16x1 male threaded adapters including a pair of 8 and 12 mm hose barbs (pump connections M16x1 male).USB connection for external PC control and data-logging. A space-saving cooling coil design yields more usable space in the bath tank. Bath lid and drain tap included.PID temperature control provides ±0.03 °C stability. Centrifugal pump capacity of 15 L/min with pressure to 5.1 psi.Stainless steel bath tank. Class III (FL) according to DIN 12876-1
Catalog No. 50-137-4372
Only null left
Add to Cart
Request a Quote
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov