Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD1.1 Monoclonal Antibody (CB3), FITC
GREENER_CHOICE

Catalog No. PIMA528765
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
PIMA528765 1 mL
1 options

Catalog No. PIMA528765

Supplier: Invitrogen™ MA528765

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD1.1 Monoclonal Antibody for Flow

CD1.1 molecules, like MHC I and II, play an important role in the immune system by presenting lipid, glycolipid and lipopeptide antigen to T and NKT cells. The chicken CD1 gene is located in the chicken MHC B locus and has an important implication for the primordial MHC gene evolution. Two proteins have been identified - CD1.1 and CD1.2. Analysis of RNA from blood cells showed that both genes are expressed in Bu-1+ cells and CD8 alpha+ cells. CD1.2 is expressed in TCR gamma delta cells but is nearly undetectable in TCR alpha beta. The monoclonal antibody CB3 antibody can identify cells of the B cell lineage and recognizes CD1.1 but not CD1.2 on the surface of transfected mammalian cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD1.1
Applications Flow Cytometry
Classification Monoclonal
Clone CB3
Concentration 0.5 mg/mL
Conjugate FITC
Formulation PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.4
Gene CD1B
Gene Accession No. 0
Gene Alias a polypeptide; CD1.1; CD1.2; CD1.2 antigen; CD1-2 antigen; CD1A; CD1A antigen, a polypeptide; CD1a molecule; CD1A2; CD1B; CD1b molecule; CD1c; chCD1-2; T-cell surface glycoprotein CD1A2 antigen; T-cell surface glycoprotein CD1b
Gene Symbols CD1B
Host Species Mouse
Immunogen Lymphocytes from the bursa of Fabricius of outbred chickens.
Purification Method purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 425802
Target Species Chicken
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.