Learn More
Medchemexpress LLC HY-P7774 10ug Medchemexpress, CD100, His (HEK293-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP777410UG
Encompass_Preferred
Medchemexpress, HY-P7774 10ug CD100, His (HEK293-expressed), Human CAS: Recombinant Human CD100, His (HEK293-expressed) is a protein from the semaphorin family with important roles in the vascular, nervous and immune systems. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3180
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.