Learn More
Invitrogen™ CD100 Polyclonal Antibody

Description
PA1-4202 detects CD100 in human samples. PA1-4202 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~158 kDa protein representing CD100 in transfected CHO cells. The PA1- 4202 immunogen is a synthetic peptide corresponding to residues K(777) P K S D F C D R E Q S(788) of human CD100.
Specifications
Specifications
| Antigen | CD100 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Sema4d |
| Gene Accession No. | Q92854 |
| Gene Alias | A8; BB18; C9orf164; CD100; coll-4; GR3; M-Sema G; M-sema-G; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4D; sema J; Sema4d; Semacl2; SEMAJ; semaphorin 4D; semaphorin H; semaphorin-4D; Semaphorin-C-like 2; semaphorin-J; Semcl2 |
| Gene Symbols | Sema4d |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.