Learn More
Invitrogen™ CD100 Polyclonal Antibody

Description
In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) Semaphorin 3A, rhSemaphorin 4B, and rhSemaphrin 6C is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | CD100 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Sema4d |
| Gene Accession No. | O09126 |
| Gene Alias | A8; BB18; C9orf164; CD100; coll-4; GR3; M-Sema G; M-sema-G; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4D; sema J; Sema4d; Semacl2; SEMAJ; semaphorin 4D; semaphorin H; semaphorin-4D; Semaphorin-C-like 2; semaphorin-J; Semcl2 |
| Gene Symbols | Sema4d |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.