Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD100 (SEMA4D) Monoclonal Antibody (eBio133-1C6 (133-1C6)), eBioscience™

Catalog No. 14100937
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-100-937 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-100-937 Supplier Invitrogen™ Supplier No. 14100937
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The eBio133-1C6 monoclonal antibody reacts with human CD100, a member of the class IV semaphorin family also called sema4D. CD100 promotes angiogenesis, invasive growth, proliferation, and anti-apoptosis of cancer cells in vitro. Furthermore CD100 is a costimulatory molecule in adaptive immune responses by switching off negative signalling from CD72. In addition to CD72 another ligand is Plexin-B1. It has been shown that higher expression levels of CD100 are correlated with poor survival in soft tissue sarcoma patients. Expression is found in both hematopoietic and non-hematopoietic cells.CD100 is expressed on resting T cells and NK cells and B cell. CD100 can also exist as a soluble form. Applications Reported: This eBio133-1C6 (133-1C6) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and ELISA. Applications Tested: This eBio133-1C6 (133-1C6) antibody has been tested by flow cytometric analysis of human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm pos...

Semaphorins are neuronal chemorepellants that direct pioneering neurons during nervous system development. CD100 (also known as semaphorin 4d), a 150 kDa homodimer, is a novel semaphorin that induces B cells to aggregate and improves their viability in vitro. CD100 modifies CD40-CD40L B cell signaling by augmenting B cell aggregation and survival and down regulating CD23 expression. CD72 is a lymphocyte receptor for the class IV semaphorin CD100, which is involved in regulating B cell signaling. CD100 is expressed on resting and PHA stimulated T cells. The protein is weakly expressed on NK cells, EBV transformed B cells, monocytes and tumor/peripheral blood T cell lines, and at higher density on activated T cells and B cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD100 (SEMA4D)
Applications ELISA, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone eBio133-1C6 (133-1C6)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Sema4d
Gene Accession No. Q92854
Gene Alias A8; BB18; C9orf164; CD100; coll-4; GR3; M-Sema G; M-sema-G; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4D; sema J; Sema4d; Semacl2; SEMAJ; semaphorin 4D; semaphorin H; semaphorin-4D; Semaphorin-C-like 2; semaphorin-J; Semcl2
Gene Symbols Sema4d
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10507
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.