Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD102 (ICAM-2) Monoclonal Antibody (CBR-IC2/2), eBioscience™

Catalog No. p-7168106
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-102-937 2 mg
50-127-89 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-102-937 Supplier Invitrogen™ Supplier No. 14102937
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The CBRIC2/2 monoclonal antibody reacts with human ICAM-2 (CD102). ICAM-2 is a 55-65 kD transmembrane glycoprotein possessing 2 extracellular Ig domains, a single transmembrane domain, and a short 26-amino acid cytoplasmic domain. ICAM-2 is expressed on most leukocytes, and is strongly expressed on vascular endothelial cells. ICAM-2 is a ligand of integrin CD11a/CD18 (LFA-1), and also has been reported to bind integrin CD11b/CD18 (Mac-1). ICAM-2 is an intercellular adhesion molecule involved in lymphocyte homing to sites of inflammation. The CBRIC2/2 antibody has been reported to have blocking activity. Applications Reported: This CBRIC2/2 antibody has been reported for use in flow cytometric analysis, immunoblotting, immunoprecipitation, immunohistochemical staining, and functional assays. Applications Tested: This CBRIC2/2 antibody has been tested by flow cytometric analysis of human peripheral blood cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. The CBRIC2/2 antibody has also been found useful for immunoblotting of a human peripheral blood cell lysate, recognizing a protein of approximately 55 kD under non-reducing conditions. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. P...

ICAM-2 is a member of the intercellular adhesion molecule (ICAM) family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. ICAM-2 may play a role in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion. It mediates adhesive interactions important for antigen-specific immune response, NK-cell mediated clearance, lymphocyte recirculation, and other cellular interactions important for immune response and surveillance. Several transcript variants encoding the same protein have been found for this gene. Diseases associated with ICAM2 include Colon Carcinoma In Situ.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD102 (ICAM-2)
Applications Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CBR-IC2/2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene ICAM2
Gene Accession No. P13598
Gene Alias CD102; Icam2; ICAM-2; Intercellular adhesion molecule 2; Ly-60; Lymphocyte function-associated AG-1 counter-receptor; RP23-418O11.10-001
Gene Symbols ICAM2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3384
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.