Learn More
Invitrogen™ CD103 (Integrin alpha E) Polyclonal Antibody

Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GGAPFVLSSL LHQDPSTNQT WLLVTSPRTK RTPGPLHRCS LVQDEILCHP VEHVPIPKGR HRGVTVVRSH HGVLICIQVL VRRPHSLSSE LTGTCSLLGP DLRPQAQANF FDLENLLDPD ARVDTGDCYS NK Highest antigen sequence indentity to the following orthologs: Mouse - 60%, Rat - 60%.
Specifications
Specifications
| Antigen | CD103 (Integrin alpha E) |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Itgae |
| Gene Accession No. | P38570 |
| Gene Alias | A530055J10; alpha-E1; alpha-M290; aM290; antigen CD103; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; CD103; DC; DCs; HML-1 antigen; human mucosal lymphocyte antigen 1, alpha polypeptide; HUMINAE; integrin alpha E; integrin alpha E, epithelial-associated; integrin alpha E1, epithelial-associated; integrin alpha M290; Integrin alpha-E; Integrin alpha-E heavy chain; Integrin alpha-E light chain; Integrin alpha-IEL; integrin subunit alpha E; integrin, alpha E; integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide); integrin, alpha E, epithelial-associated; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen |
| Gene Symbols | Itgae |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.