Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD104 Rat anti-Mouse, Brilliant Violet 421, Clone: 346-11A, BD Optibuild™

Catalog No. BDB743078 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743078 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743078 Supplier BD Biosciences Supplier No. 743078
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 346-11A monoclonal antibody specifically recognizesan epitope at the beginning of the cysteine-rich repeat region of the 200-kDa integrin β4 chain (CD104), which is found on the cell surface as a heterodimeric complex with the integrin α6 chain (CD49f). The α6β4(CD49f/CD104) complex binds to laminins and is expressed on the basal surface of a variety of epithelial cell types, particularly on stratified squamous epithelia, and is also found in peripheral nerves, in certain subsets of endothelial cells, and on immature thymocytes. It has also been identified on a number of tumor tissues and participates in tumor progression events. Localization of both human and mouse epidermal α6β4 integrin to hemidesmosomes suggests that this heterodimer playsa role in epidermal adhesion to the basement membrane.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD104
Classification Monoclonal
Clone 346-11A
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias β4 integrin, Integrin β4 chain, Integrin beta-4, Itgb4, ITB4
Host Species Rat
Immunogen Tumor-associated antigen TSP-180 from a BALB/c mouse lung carcinoma
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.