Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD105 (Endoglin) Monoclonal Antibody (SN6), Brilliant Violet™ 605, eBioscience™
GREENER_CHOICE

Catalog No. 406105742
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-610-5742 100 Tests
40-610-5741 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-610-5742 Supplier Invitrogen™ Supplier No. 406105742
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

The SN6 monoclonal antibody reacts with human CD105, also known as Endoglin. CD105, an approximately 90 kDa disulfide-linked homodimer is expressed by vascular endothelial cells and some bone marrow cells and activated macrophages. It is suggested that CD105 functions in adhesion and embryonic angiogenesis. Applications Tested This SN6 antibody has been pre-diluted and tested by flow cytometric analysis of U937 cell line. This may be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Blocking Buffers When using two or more Super Bright, Brilliant Violet™, Brilliant Ultra Violet™, or other polymer dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) or Brilliant Stain Buffer (Product # 00-4409-75) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer or Brilliant Stain Buffer for more information. Light sensitivity This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step...

CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD105 (Endoglin)
Applications Flow Cytometry
Classification Monoclonal
Clone SN6
Concentration 5 μL/Test
Conjugate Brilliant Violet 605
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene ENG
Gene Accession No. P17813
Gene Alias AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein
Gene Symbols ENG
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2022
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.