Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD105 Monoclonal Antibody (MEM-226), Alexa Fluor™ 488
GREENER_CHOICE

Catalog No. PIMA518120 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518120 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518120 Supplier Invitrogen™ Supplier No. MA518120
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with an extracellular epitope of CD105 (Endoglin), a 90 kDa type I homodimerizing membrane glycoprotein expressed on vascular endothelial cells (small and large vessels), activated monocytes and tissue macrophages, stromal cells of certain tissues including bone marrow, pre-B lymphocytes in fetal marrow and erythroid precursors in fetal and adult bone marrow; it is also present on syncytiotrophoblast on placenta throughout pregnancy.

CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD105
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone MEM-226
Concentration 0.055 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS with 0.2% BSA and 15mM sodium azide
Gene ENG
Gene Accession No. P17813
Gene Alias AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein
Gene Symbols ENG
Host Species Mouse
Immunogen Recombinant Vaccinia virus containing the human CD105 cDNA.
Purification Method Size-exclusion chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2022, 497010
Target Species Human, Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.