Learn More
Epredia™ Lab Vision™ CD106/VCAM-1 Ab-3 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: 1.4C3
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Stimulated human umbilical vein endothelial cells (HUVEC)
Molecular Weight: 110 kDa
Positive Control: Placenta
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Frozen and formalin/paraffin)
- Western Blotting (Not recommended)
Note:
USA: RUO; Int'l: RUO
General References:1. Thornhill M, et. al. (1990) J Immunol, 145:865.
2. Wellicome s M, et. al. (1990) J Immunol, 144:2558.
3. Kyan-Aung U, et. al. (1991) J Immunol, 146:521-528
4. Burrows f J, et. al. Cancer Res, 51:4768.
5. Osborne L, et al. (1994) J Cell Biol, 124:601.
6. Thornhill M, et. al. (1991) J Immunol, 146:592.
Specifications
Specifications
| Antigen | CD106/VCAM-1 Ab-3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 1.4C3 |
| Conjugate | Unconjugated |
| Formulation | Ready-to-use for IHC (UVLP) |
| Host Species | Mouse |
| Immunogen | Stimulated human umbilical vein endothelial cells (HUVEC) |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.