Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD106 (VCAM-1) Monoclonal Antibody (B-N8), eBioscience™
GREENER_CHOICE

Catalog No. 5017797
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-177-97 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-177-97 Supplier Invitrogen™ Supplier No. BMS141
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VCAM-1 (B-N8) is a mouse monoclonal antibody raised against VCAM-1 ofhuman origin. B-N8 can be used for functional studies of cell adhesion molecules. BMS141BT can be used for FACS analysis as determined using stimulated human umbilical vein endothelial cells (HUVEC). The antibody can be used to stain aceton-fixed cryostat sections. BMS141 is suitable for ELISA technique. Applications Tested: ELISA, Flow Cytometry, Functional Studies, Immunohistochemistry. Purity: >95%.

VCAM-1 is a cell surface sialoglycoprotein expressed by cytokine activated endothelium. The protein has a number of functions including the regulation of leukocyte migration, leukocyte-endothelial cell adhesion and signal transduction and may play a role in a number of inflammatory diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD106 (VCAM-1)
Applications ELISA, Flow Cytometry, Functional Assay, Immunohistochemistry
Classification Monoclonal
Clone B-N8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene VCAM1
Gene Accession No. P19320
Gene Alias adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B
Gene Symbols VCAM1
Host Species Mouse
Immunogen Rec. Human VCAM-1.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7412
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.