Learn More
MBL International Corporation IMMUNOCYTO CD107a Detection Kit
SDP
Supplier:  MBL International Corporation 4844
Encompass_Preferred
Size:50 Assays; The IMMUNOCYTO CD107a Detection Kit is a reagent for detection of CD107a exposed on the cell surface after antigen stimulation by CD107a mobilization assay.IMMUNOCYTO CD107a Detection Kit is intended for the detection of CD107a in research samples.
Catalog No. 50-446-23
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.