Learn More
Invitrogen™ CD107a (LAMP-1) Monoclonal Antibody (eBioH4A3), Brilliant Violet™ 650, eBioscience™, Invitrogen™

Description
Specifications
Specifications
| Antigen | CD107a (LAMP-1) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | eBioH4A3 |
| Concentration | 5 μL/Test |
| Conjugate | Brilliant Violet 650 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | LAMP1 |
| Gene Accession No. | P11279 |
| Gene Alias | 120 kDa lysosomal membrane glycoprotein; AI196048; CD107 antigen-like family member A; CD107a; I79_011073; Lamp I; LAMP1; LAMP-1; LAMPA; LGP120; LGP-120; LGPA; LGP-A; lysosomal associated membrane protein 1; Lysosomal associated membrane protein 1 (120 kDa); lysosomal membrane glycoprotein 1; lysosomal membrane glycoprotein A; lysosomal-associated membrane protein 1; lysosome-associated membrane glycoprotein 1; LYSOSOME-ASSOCIATED MEMBRANE GLYCOPROTEIN 1 PRECURSOR (LAMP-1) (LGP-A) (LGP-120) (CD107A) (P2B); lysosome-associated membrane protein 1; P2B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.