Learn More
Description
Specifications
Specifications
| Antigen | CD107a |
| Applications | Flow Cytometry, Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 1D4B |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Ultraviolet 395 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | LAMP-1; LGP-120; LGP-A; P2B |
| Host Species | Rat |
| Immunogen | Plasma membrane fraction of mouse embryo NIH 3T3 cell line |
| Show More |
For Reseach Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.