Learn More
Invitrogen™ CD107b (LAMP-2) Monoclonal Antibody (eBioABL-93 (ABL-93)), eFluor™ 660, eBioscience™

Description
The eBioABL-93 monoclonal antibody reacts with mouse CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated protein of approximately 120 kDa that is expressed intracellularly in lysosomal membranes. It is also transiently expressed on the surface of cytolytic T cells during degranulation, but to a lesser extent than CD107a. It has been reported that CD107b may be identical to the mouse Mac-3 antigen. Applications Reported: This ABL-93 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This eBioABL-93 (ABL-93) antibody has been tested by intracellular staining and flow cytometric analysis of thioglycolate elicited peritoneal mouse cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD107b (LAMP-2) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | eBioABL-93 (ABL-93) |
| Concentration | 0.2 mg/mL |
| Conjugate | eFluor 660 |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | LAMP2 |
| Gene Accession No. | P17047 |
| Gene Alias | CD107 antigen-like family member B; CD107b; EGK_20887; Igp96; Lamp II; LAMP2; LAMP-2; LAMP-2 Isoform 2A; LAMP-2 Variant A; Lamp-2a; Lamp-2b; Lamp-2c; LAMPB; LGP110; LGP-110; LGP-96; LGP-B; lysosomal associated membrane protein 2; lysosomal membrane glycoprotein 2; lysosomal membrane glycoprotein type B; lysosomal-associated membrane protein 2; lysosome-associated membrane glycoprotein 2; lysosome-associated membrane protein 2; Mac3; RP23-193O17.2 |
| Show More |
Frequently Asked Questions (FAQs)
Yes, in-house studies have demonstrated that the eFluor 660 fluorochrome is recognized by Anti-Cy5/Alexa Fluor 647 beads. Side by side studies with Alexa Fluor 647 versus eFluor 660 conjugated antibodies have demonstrated comparable results.
The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.
The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.
As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.