Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD107b (LAMP-2) Monoclonal Antibody (eBioH4B4 (H4B4)), eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5012817
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-128-17 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-128-17 Supplier Invitrogen™ Supplier No. 14107882
  • $161.00
    $120.65 / Each of 1

    Save $40.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The eBioH4B4 monoclonal antibody reacts with human CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated, type I transmembrane protein of approximately 105 kDa. It is expressed intracellularly in lysosomal/endosomal membranes in nearly all cells. It is also expressed on the surface of degranulating T cells (to a lesser extent than CD107a) and activated platelets as well as some cancer cells. In humans, mutations in CD107b results in a lysosomal glycogen storage disorder, known as Danon disease. Applications Reported: Purified anti-human CD107b (LAMP-2) has been reported for use in flow cytometric analysis. It has also been reported for use in surface staining in a flow cytometric based degranulation assay. (Fluorochrome conjugated eBioH4B4 (H4B4) is recommended for use in flow cytometry.). Applications Tested: This eBioH4B4 (H4B4) antibody has been tested by intracellular staining and flow cytometric analysis of Jurkat cell line. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. This antibody has also been tested by immunoblotting at 1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

LAMP2 (Lysosome-associated membrane glycoprotein 2, CD107b) is a member of a family of membrane glycoproteins. LAMP2 provides selectins with carbohydrate ligands, and may play a role in tumor cell metastasis, and have a function in the protection, maintenance, and adhesion of the lysosome. Prior to posttranslational modification, LAMP2 is approximately 46 kDa polypeptide. Mature, functional LAMP2 is extensively glycosylated with a variety of different N-linked and O-linked oligosaccharides with a total molecular weight of about 100-130 kDa. LAMP 2 has a structure comprised of a large amino-terminal intra-lysosomal domain, a hydrophobic trans-membrane domain, and a short carboxyl-terminal cytoplasmic tail. LAMP2 is used to monitor a very selective form of autophagy called chaperone-mediated autophagy. LAMP 2 has a structure comprised of a large amino-terminal intra-lysosomal domain, a hydrophobic trans-membrane domain, and a short carboxyl-terminal cytoplasmic tail. The LAMP protein has 3 variant forms: LAMP-2A, -2B, and 2C. LAMP2, together with CD107a/LAMP-1, is a major constituent of lysosomal membrane. The LAMP proteins are involved in lysosome biogenesis and are required for fusion of lysosomes with phagosomes, and LAMP2 is an important regulator in successful phagosomal maturation. LAMP2 deficiency causes an accumulation of autophagosomes in many tissues leading to cardiomyopathy and myopathy (Danon's disease).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD107b (LAMP-2)
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone eBioH4B4 (H4B4)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene LAMP2
Gene Accession No. P13473
Gene Alias CD107 antigen-like family member B; CD107b; EGK_20887; Igp96; Lamp II; LAMP2; LAMP-2; LAMP-2 Isoform 2A; LAMP-2 Variant A; Lamp-2a; Lamp-2b; Lamp-2c; LAMPB; LGP110; LGP-110; LGP-96; LGP-B; lysosomal associated membrane protein 2; lysosomal membrane glycoprotein 2; lysosomal membrane glycoprotein type B; lysosomal-associated membrane protein 2; lysosome-associated membrane glycoprotein 2; lysosome-associated membrane protein 2; Mac3; RP23-193O17.2
Gene Symbols LAMP2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3920
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.